Learn More
Fisherbrand™ Sterile Graduated Transfer Pipets
Made up of low density polyethylene.
Brand:  Fisherbrand™ 13409118
 View more versions of this product
Product Code. 13409118
  • 31.80 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Transparent in color
  • Contains graduations
  • Sterile
Specifications
Specifications
LDPE
Yes
500/Cs.
25 inner packs of 20
155 mm
25
0.5 mL
40 μL
3 mL
Transfer Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Sterile Graduated Transfer Pipets > Capacity: 3mL; Drops/mL: 25/ 20 Case