missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ Sterile HydroPure
Leaves little to no residue and has a guaranteed endotoxin level of less than 0.25 EU/mL making it ideal for use in product contact areas. Contec™ Sterile HydroPure is a blend of hydrogen peroxide and water for injection or purified water.
Brand: Contec™ SBC16HP
184.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hydrogen peroxide breaks down to water and oxygen making it very low residue and suitable for product contact areas
- Guaranteed endotoxin level of less than 0.25 EU/mL so ideal for use in product contact area
- Sterile filtered to 0.2 microns under Grade A airflow in a Grade B cleanroom into pre-irradiated containers
- Sporicidal in 15 mins so has sufficient sporicidal activity for a Grade A / B areas
Specifications
Sporicidal Disinfectant and Cleaner | |
Not Applicable | |
Clear | |
Grade A / B Areas | |
Fluid | |
Capped Bottle | |
Filtered | |
1 L |
Liquid | |
Sterile | |
Not Autoclavable | |
Clear | |
6% Hydrogen Peroxide in Purified Water | |
Limited-use | |
6 x 1 L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.