Learn More
Pro-Lab Diagnostics™ Sterile Inoculation Loop
For inoculation with nichrome wire loops. Prolab Diagnostics™ Sterile Inoculation Loops are part of a complete range of inoculation tools including traditional calibrated nichrome wire loops with a selection of insulated holders.
Brand: Pro-Lab Diagnostics™ PL-101
This item is not returnable.
View return policy
Description
- Accurately calibrated nichrome wire loops
- Standard gauge
- Supplied in packs of 25
Specifications
5 μL | |
Nichrome | |
Inoculating Loop | |
25 Pk. | |
Laboratory and Research use |
3.57 mm | |
Sterile | |
Laboratory and Research use | |
Yes |