Learn More
Gammex™ Sterile Latex Surgical Gloves Product Code.: 13154724
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
Gammex™ Sterile Latex Surgical Gloves
Product Code. 13154724
6
Click to view available options
6
Product Code. 13154724
Brand: Gammex™ AJ330048060
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
Add to basket
Gammex™ Sterile Latex Surgical Gloves are advanced powder-free latex gloves for general surgery with a micro-rough outer surface for enhanced grip.
  • Advanced powder-free latex surgical gloves
  • Optimal combination of safety and comfort
  • Provides a secure barrier without compromising sensitivity
  • Donning is made easy, even with damp hands, thanks to the water-based polyurethane inner coating
  • Micro-rough outer surface for enhanced grip
  • Very low latex protein and chemical accelerator residue content safe on skin
  • For laboratory research purposes only
Specifications
Certifications/Compliance AS/NZS 4179, ASTM F739, EN 374-1/2/3, EN 420, EN 455 1-4, ISO 10282, JIS T9107
Color Natural
Disposable Single-use
Hand Ambidextrous
Latex Free No
Material Latex
Powdered Powder Free
Size 6
Sterility Sterile
Glove Cuff Style Straight
Product Content Correction
Product Title
Gammex™ Sterile Latex Surgical Gloves >