missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile IPA
BIOPHARMA

Provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.

230.00 € - 242.00 €

Specifications

pH Not Applicable
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
View More Specs

Products 3
Product Code Brand Container Type Quantity Price Quantity & Availability  
Product Code Brand Container Type Quantity Price Quantity & Availability  
15892917
BIOPHARMA
View Documents
Contec™
SBT0570IW
Trigger Spray Bottle 8 x 0.5 L
242.00 €
8 bottles
Please to purchase this item. Need a web account? Register with us today!
15597171
BIOPHARMA
View Documents
Contec™
SBT170IW
Trigger Spray Bottle 6 x 1 L
235.00 €
6 bottles
Please to purchase this item. Need a web account? Register with us today!
15537171
BIOPHARMA
View Documents
Contec™
SBC570I
Capped Bottle 2 x 5 L
230.00 €
2 bottles
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
  • Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
Specifications

Specifications

Not Applicable
Colorless
Clear
Filtered
Sterile
Not Autoclavable
Limited-use
Yes
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CONTEC STERILE 70% IPA WITH WFI

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : PROPAN-2-OL

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.