Learn More
Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase performance through the superior tip straightness and low %CV with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems.
Brand: Thermo Scientific™ 965-261
375.08 EUR valid until 2024-12-31
Use promo code "21899" to get your promotional price.
Code : 04
Additional Details : Weight : 7.35000kg
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:- Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384-, and 1536-deep well plates
- Low percentage coefficient of variation (%CV)
Specifications
30 μL | |
Sterile | |
Beckman Coulter Liquid Handling Systems |
Filtered | |
Extended Length |