missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Sterile Plastic Inoculation Loops (Volume: 1μL)

Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation.

Brand:  Microspec™ C4G

 View more versions of this product

Product Code. 15792115

  • 55.05 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 1μL
  • Color: Blue
Specifications

Specifications

Single-use
0.04 in.
7.87 in.
High-impact Polystyrene
Blue
7.87 in.
Microseal Pouch
1 μL
1 mm
200 mm
Gamma-irradiated
Plastic Culture Loops
50 Pouches of 20
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers