missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 10μL)
Retrieves an inoculum from culture of microorganisms.
50.30 € - 520.00 €
Specifications
Volume | 10 μL |
---|---|
Color | Blue |
Length (English) Handle | 7.87 in. |
Length (Metric) Handle | 200 mm |
Product Code | Brand | Quantity | Packaging Type | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Sterility | Price | Quantity & Availability | |||
15702115
|
Microspec™
A4G |
50 Pouches of 20 | Microseal Pouch | Gamma-irradiated |
61.65 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||
15772105
|
Microspec™
A1 |
16000 | Plastic Vial | Non-sterile | N/A | ||||
15782105
|
Microspec™
A1G |
800 Pouches of 20 | Microseal Pouch | Sterile | N/A | ||||
15792105
|
Microspec™
A4 |
50 Bags of 20 | Microseal Pouch | Aseptically Manufactured |
50.30 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 10μL
- Color: Blue
Specifications
10 μL | |
Blue | |
7.87 in. | |
200 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title