missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Sterile Plastic Inoculation Loops (Volume: 5μL)

Product Code. 15742115
Click to view available options
Quantity:
50 Pouches of 20
800 Pouches of 20
Packaging Type:
Grip Seal Bag
Microseal Pouch
Sterility:
Aseptically Manufactured
Gamma-irradiated
Sterile
Unit Size:
Pack of 1000
Pack of 16000

Product Code. 15742115

Brand: Microspec™ B4

Only null left
Add to basket
Edge
Add to basket

Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 5μL
  • Color: Blue

Specifications

Disposable Single-use
Diameter (English) Inside 0.10 in.
Diameter (Metric) Inside 2.5 mm
Length (English) 7.87 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Aseptically Manufactured
Color Blue
Product Type Inoculating Product
Length (English) Handle 7.87 in.
Length (Metric) Handle 200 mm
Quantity 50 Pouches of 20
Packaging Type Microseal Pouch
Calibrated Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.