Learn More
Fisherbrand™ Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Pipets have improved optical clarity — high-contrast black markings make volumes easier to read
57.10 € - 286.00 €
Specifications
Certifications/Compliance | Non-pyrogenic, USP XXII, USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free |
---|---|
Material | Polystyrene |
Graduated | Yes |
Disposable | Single-use |
Product Code | Brand | Volume (Metric) | Packaging Type | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Packaging Type | Tolerance | Price | Quantity & Availability | |||
11849181
![]() |
Fisherbrand™
431028 |
1 mL | Individual Paper/Plastic Wrap | ±0.02 mL |
173.00 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
11819660
![]() |
Fisherbrand™
4311 |
1 mL | Individual All Plastic Wrap | ±0.02 mL |
209.00 €
Pack of 1000 |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||
11889650
![]() |
Fisherbrand™
4313 |
1 mL | Bulk | ±0.02 mL |
252.00 €
Pack of 1000 |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||
11859181
![]() |
Fisherbrand™
431029 |
2 mL | Individual Paper/Plastic Wrap | ±0.04 mL |
99.20 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
11879650
![]() |
Fisherbrand™
4320 |
2 mL | Individual All Plastic Wrap | ±0.04 mL |
67.70 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
11869650
![]() |
Fisherbrand™
4321 |
2 mL | Bulk | ±0.04 mL |
76.99 €
Pack of 500 |
In Stock
Log in to see stock. |
|||
11869181
![]() |
Fisherbrand™
431030 |
5 mL | Individual Paper/Plastic Wrap | ±0.1 mL |
57.10 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11829660
![]() |
Fisherbrand™
4330 |
5 mL | Individual All Plastic Wrap | ±0.1 mL |
58.60 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11899650
![]() |
Fisherbrand™
4331 |
5 mL | Bulk | ±0.1 mL |
149.00 €
Pack of 500 |
In Stock
Log in to see stock. |
|||
11879181
![]() |
Fisherbrand™
431031 |
10 mL | Individual Paper/Plastic Wrap | ±0.2 mL |
88.80 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11839660
![]() |
Fisherbrand™
4340 |
10 mL | Individual All Plastic Wrap | ±0.2 mL |
59.20 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11809660
![]() |
Fisherbrand™
4341 |
10 mL | Bulk | ±0.02 mL |
149.00 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
11839181
![]() |
Fisherbrand™
431032 |
25 mL | Individual Paper/Plastic Wrap | ±0.5 mL |
145.23 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11517752
![]() |
Fisherbrand™
4350 |
25 mL | Individual All Plastic Wrap | ±0.5 mL |
128.96 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
12559546
![]() |
Fisherbrand™
4351 |
25 mL | Bulk | ±0.5 mL |
128.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11889181
![]() |
Fisherbrand™
431033 |
50 mL | Individual Paper/Plastic Wrap | ±1 mL |
196.56 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
11537752
![]() |
Fisherbrand™
4360 |
50 mL | Individual All Plastic Wrap | ±1 mL |
164.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
11859660
![]() |
Fisherbrand™
4361 |
50 mL | Bulk | ±1 mL |
146.00 €
Pack of 100 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||
16348745
![]() |
Fisherbrand™
4400 |
100 mL | Individual Paper/Plastic Wrap | ±2% |
286.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
16338745
![]() |
Fisherbrand™
431052 |
100 mL | Individual All Plastic Wrap | ±2% |
264.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
The high-contrast black markings make the volumes easier to read, for improved optical clarity. Manufactured with optically clear polystyrene for accurate measurement. Fisherbrand™ Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe reduce reading errors and are designed for tissue culture work.
- Color coded magnifier stripe runs lengthwise along pipet, making meniscus obvious at arm's length — reducing reading errors
- Sterile and nonwettable pipets
- Offer a tolerance of ±2% at full volume and have plug-in mouth end
- 5, 10, 25, and 50mL pipets have ascending and descending graduations
- Improved tip design on 25 and 50mL pipets increases the effect of surface tension to minimize dripping
- Color coding on each pipet and on inner bags for multipack pipets make it easy to spot the size needed
- Each pipet lot is tested for sterility, accuracy and endotoxin level
- Bulk pack and individually wrapped, with paper and plastic wraps
- Cotton plugged
Order Info
Inner bags are recyclable LDPE; outer boxes also are recyclable. Individually wrapped pipets are ideal for small-volume use requiring sterility and are available in either a clear, durable all-plastic wrap, or a color-coded paper/plastic wrap for reduced static cling. Bulk pack for larger-volume use permits easy access to several pipets at a time without contaminating unused pipets.
Certifications
Certified nonpyrogenic with an endotoxin level less than 0.5EU/mL following USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free
Compliance
Calibrated To Deliver/Blow Out
Specifications
Non-pyrogenic, USP XXII, USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free | |
Polystyrene | |
Yes | |
Single-use |