missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Service Consultants™ Sterile Polystyrene L-shaped Spreader
Click to view available options
Color:
Blue
Clear
Description:
Firm Plastic
Soft Plastic
Unit Size:
Pack of 1000
Description
- Ideal for bacterial counts
- General laboratory uses for sample throughput or protect
- Sterilized by gamma irradiation; sterility guaranteed only while bag is sealed
- Shelf life: 36 months from date of manufacture
- Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Specifications
| Description | Firm Plastic |
| Color | Clear |
| Quantity | 1000/Pk. |
| Shape | L-Shape |
| Material | Polystyrene |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction