missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Autoclave Bags
GREENER_CHOICE

Product Code. 11379103
Click to view available options
Dimensions (L x W):
601 x 406mm
610 x 406mm
660 x 305mm
810 x 610mm
Material:
HDPE
Polypropylene
No. per Case:
200
500
Unit Size:
Pack of 200
Pack of 500

Product Code. 11379103

Brand: Thermo Scientific™ 509HT

Only null left
Add to basket
Edge
Add to basket

Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags

Ordering Alerts:

Not available in all geographic regions

TRUSTED_SUSTAINABILITY

Specifications

Description Autoclave bags (PP)
Material Polypropylene
No. per Case 200
Temperature Range (Metric) 135°C
Dimensions (L x W) 660 x 305mm
Length (English) 26 in.
Length (Metric) 660 mm
Width (English) 12 in.
Width (Metric) 305 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.