Learn More
Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
Brand: Thermo Scientific™ 612F96
86.50 EUR valid until 2025-12-31
Use promo code "24284" to get your promotional price.
Description
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.
- Manufactured from virgin polystyrene, giving excellent optical clarity
- Aseptically manufactured plates receive no surface treatment and therefore provide a medium binding surface
- Irradiation process for the sterile plates makes the surface more hydrophilic, therefore generating a high binding product
- Suitable for use at a wavelength of 340nm
- Condensation rings on the lid, together with raised well rims on the base, help minimize the risk of contamination from surrounding wells
- Orientation corners and alphanumeric labeling ensure easy sample identification
- Frosted write-on area on end wall of plate for clear identification
- Hanging well design ensures even temperature distribution around each well
- The plates are supplied without a lid (can be purchased separately)
- In vitro use only
Ordering Alerts:
Not available in all geographic regions
Specifications
Standard | |
8 x 12 | |
Clear | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
1 |
Microtiter Plates, Clear, Flat bottom | |
50 Cases | |
Microplate | |
96 | |
Non-treated | |
50 | |
50/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.