Learn More
Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
Brand: Thermo Scientific™ 642000
49.81 EUR valid until 2025-03-14
Use promo code "24150" to get your promotional price.
Description
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.
Features
- Manufactured from virgin polystyrene for excellent optical clarity
- Aseptically manufactured without surface treatment for a medium binding surface
- Irradiated for sterility and a more hydrophilic surface, providing high binding
- Suitable for use at a wavelength of 340nm
- Minimizes the risk of contamination from surrounding wells with condensation rings on the lid raised well rims on the base.
- Ensures even temperature distribution around each well with a hanging well design
Easy Sample Identification
- Frosted writing area on the end wall
- Orientation corners
- Alphanumeric labeling
Note:
For in vitro use only. Plates are supplied without a lid (can be purchased separately).
Ordering Information:
Product is not available in all geographic regions.

Specifications
50 Cases, 1 Pk. | |
Plate Lid | |
96 | |
High Binding | |
50 |
Clear | |
Polystyrene | |
Sterile | |
Not Applicable | |
1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.