missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates

Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.

Brand:  Thermo Scientific™ 611U96

41.18 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.


 View more versions of this product

Product Code. 11339163

  • 61.15 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.

  • Manufactured from virgin polystyrene, giving excellent optical clarity
  • Aseptically manufactured plates receive no surface treatment and therefore provide a medium binding surface
  • Irradiation process for the sterile plates makes the surface more hydrophilic, therefore generating a high binding product
  • Suitable for use at a wavelength of 340nm
  • Condensation rings on the lid, together with raised well rims on the base, help minimize the risk of contamination from surrounding wells
  • Orientation corners and alphanumeric labeling ensure easy sample identification
  • Frosted write-on area on end wall of plate for clear identification
  • Hanging well design ensures even temperature distribution around each well
  • The plates are supplied without a lid (can be purchased separately)
  • In vitro use only

Ordering Alerts:

Not available in all geographic regions

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Clear
Polystyrene
Non-sterile
Round Bottom (U-well)
5
Microtiter Plates, Clear, U bottom, NS
50 Cases
Microplate
96
Non-treated
50
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers