missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates

Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.

Brand:  Thermo Scientific™ 611V96

39.83 EUR valid until 2025-12-31
Use promo code "24284" to get your promotional price.


 View more versions of this product

Product Code. 11897403

  • 56.10 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.

  • Manufactured from virgin polystyrene, giving excellent optical clarity
  • Aseptically manufactured plates receive no surface treatment and therefore provide a medium binding surface
  • Irradiation process for the sterile plates makes the surface more hydrophilic, therefore generating a high binding product
  • Suitable for use at a wavelength of 340nm
  • Condensation rings on the lid, together with raised well rims on the base, help minimize the risk of contamination from surrounding wells
  • Orientation corners and alphanumeric labeling ensure easy sample identification
  • Frosted write-on area on end wall of plate for clear identification
  • Hanging well design ensures even temperature distribution around each well
  • The plates are supplied without a lid (can be purchased separately)
  • In vitro use only

Ordering Alerts:

Not available in all geographic regions

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE marked in accordance with the European Directive 98/72/EC
Microtiter Plates, Clear, V bottom
50 Cases
Microplate
96
Non-treated
50
50/Pk.
Standard
8 x 12
Clear
Polystyrene
Non-sterile
Conical Bottom (V-well)
5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.