missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sterilin™ Cuvets
Experience top optical quality in spectrophotometry with easy-to-use, polystyrene or acrylic Thermo Scientific™ Sterilin™ Cuvets.
Brand: Thermo Scientific™ 222M
This item is not returnable.
View return policy
Description
Experience top optical quality in spectrophotometry with easy-to-use, polystyrene or acrylic Thermo Scientific™ Sterilin™ Cuvets.
- Choice of material: optical quality polystyrene for use at visible wavelengths (340 to 900nm) or optical quality acrylic (PMMA) for use at ultraviolet wavelengths (300 to 900nm)
- Each case contains cuvets from the same mold cavity to ensure minimal dimensional variation
- Standard absorption variation between cuvets is better than ±0.005 absorbency units
- Two sizes available—semi-micro cuvet and macro cuvet
- Standard pathlength of 10mm
- Frosted sides on the cuvet provide an ideal labeling and handling area
- Recessed windows reduce the risk of scratching during use
- For ease of use an arrow feature indicates the direction of light transmission
- Packed in expanded polystyrene (EPS) foam trays for optimum protection ensuring scratchfree product at the time of use
- Not available in all geographic regions
Specifications
Non-sterile | |
Macro cuvet | |
PMMA | |
10 mm | |
32.5 mm | |
10 mm |
Macro Cuvette | |
2.5 to 4.5 mL | |
100 pk/ 500 cs | |
300 to 900 nm | |
Spectrophotometers and Photometers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Note: Cuvets should not be used for long-term storage of samples. When using Hydrochloric acid, as the instrument can come under attack from the acid fumes, it is recommended that you use sealing film on the cuvet.
Spot an opportunity for improvement?Share a Content Correction