Learn More
Thermo Scientific™ Sterilin™ Dippas™
Facilitate sample collection and transportation to the lab in a single container using Thermo Scientific™ Sterilin™ Dippas™ to reduce risk of cross-contamination.
179.00 € - 487.00 €
Product Code | Brand | Capacity (Metric) | Length (Metric) | Closure Material | Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Closure Material | Color | Price | Quantity & Availability | ||
11729352
|
Thermo Scientific™
191-17 |
30 mL | 191 mm | Polypropylene | Clear |
179.00 €
Pack of 50 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||
11738938
|
Thermo Scientific™
191BLUE |
30 mL | 191 mm | Polypropylene | Blue |
179.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11788848
|
Thermo Scientific™
194IW |
100 mL | 383 mm | Metal | Clear |
482.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11798848
|
Thermo Scientific™
194IWBLUE |
100 mL | 383 mm | Metal | Blue |
487.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11748938
|
Thermo Scientific™
192-17 |
250 mL | 334 mm | Metal | Clear |
307.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
11758938
|
Thermo Scientific™
192BLUE |
250 mL | 334 mm | Metal | Blue |
307.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Facilitate sample collection and transportation to the lab in a single container using Thermo Scientific™ Sterilin™ Dippas™ to reduce risk of cross-contamination.
NOT AVAILABLE IN AMERICA
- Individually wrapped and sterilized by gamma irradiation
- Leak tested in accordance with BS EN ISO 6717
- Available in either clear or blue polystyrene
- Handle neatly snaps off after sample collection for ease of transportation
- Choice of handle length to accommodate the majority of sampling applications
Specifications
Yes | |
Polystyrene | |
Screw Cap |