Learn More
Thermo Scientific™ Sterilin™ Plastic Round Base Tubes
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.
83.35 € - 950.00 €
Specifications
Description | Round Base Plastic Tubes |
---|---|
Material | Polystyrene |
Sterility | Non-sterile |
Product Code | Brand | Tubing Size | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Packaging Quantity | Price | Quantity & Availability | ||||
11748868
|
Thermo Scientific™
RT15 |
0.6 mL | 1000 |
950.00 €
Pack of 10000 |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||
13015723
|
Thermo Scientific™
RT20 |
1.0 mL | 1000 |
760.00 €
Pack of 10000 |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||
12842863
|
Thermo Scientific™
30908 |
2.5 mL | 2500 | N/A | |||||
11708988
|
Thermo Scientific™
RT25 |
2.7 mL | 500 | N/A | |||||
11778908
|
Thermo Scientific™
RT30 |
4.9 mL | 250 | N/A | |||||
12852863
|
Thermo Scientific™
30932 |
11.0 mL | 1500 | N/A | |||||
Description
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile.
Not available in all geographic regions.
- Suitable for samples ranging from 0.6 to 11mL
- Supplied nonsterile
Specifications
Round Base Plastic Tubes | |
Polystyrene | |
Non-sterile |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.