missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes
GREENER_CHOICE

Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.

60.65 € - 365.00 €

Specifications

Material LDPE
View More Specs

Products 10
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
12153390
View Documents
Thermo Scientific™
201C
1 mL 500/Pk., 3000/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11329203
View Documents
Thermo Scientific™
201C
1 mL 500/Pk., 3000/Cs.
103.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11339203
View Documents
Thermo Scientific™
PP88SA
1 mL 10/Pk., 500/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11349203
View Documents
Thermo Scientific™
PP88SB
1 mL 1/Pk., 500/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12027502
GREENER_CHOICE
View Documents
Thermo Scientific™
200C
3 mL 500/Pk., 3000/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12909487
GREENER_CHOICE
View Documents
Thermo Scientific™
200C
3 mL 500/Pk., 3000/Cs.
72.35 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11369203
View Documents
Thermo Scientific™
PP89SA
3 mL 10/Pk., 500/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11379203
View Documents
Thermo Scientific™
PP89SB
3 mL 1/Pk., 500/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12999477
GREENER_CHOICE
View Documents
Thermo Scientific™
202C
500/Pk., 3000/Cs.
60.65 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11758908
GREENER_CHOICE
View Documents
Thermo Scientific™
202C
500/Pk., 3000/Cs.
365.00 €
Pack of 3000
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
 
Description

Description

Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.

  • Manufactured from nontoxic low density polyethylene
  • Integral filling bulb eliminates contamination associated with separate rubber bulbs
  • Excellent transparency and uniform thickness
  • Precise graduations on 1 and 3mL versions ensuring consistent results
  • Can be heat sealed for sample storage and transportation
  • Heat sealed bulbs suitable for storage in the gas phase of liquid nitrogen
  • 3mL graduated: suitable for blood banking and general laboratory use
  • Narrow stem: suitable for use as a freeze vial, sedimentation and transport
  • 1mL graduated: suitable for general laboratory use, storage and transportation
  • of samples
Ordering Information
  • Not available in all geographic regions
Specifications

Specifications

LDPE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.