missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Polystyrene Containers, 60mL to 250mL
GREENER_CHOICE

Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.

Brand:  Thermo Scientific™ 165A

188.00 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 11319133

  • 566.00 € / Pack of 120

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.

NOT AVAILABLE IN AMERICA

Features:
  • Aseptically manufactured
  • Leak-tested in accordance with BS EN 14254 Annexe D
  • 125AM, 125BM and 125CM product certified 95kPa-compliant ensuring suitability for sample transport

Note:

For in vitro use only.

Ordering Information:

Product is not available in North America.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Container
Polystyrene
Non-sterile
Leak-tested in accordance with BS EN ISO 6717
55 mm
No Label
Yes
Cylindrical
150 mL
Metal
108 mm
120/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers