Learn More
Thermo Scientific™ Sterilin™ Weighing Boats
GREENER_CHOICE
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners.
Brand:  Thermo Scientific™ WB30322
 View more versions of this product
Product Code. 11844712
  • 80.50 € / Pack of 500
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners. All sizes packed 250 per pack, 500 per case.
  • Manufactured from high impact polystyrene
  • Hydrophobic surface ensures no absorption of water from atmosphere or sample
  • Available in black or white to ensure suitability for different colored sample materials
  • Anti-static range alleviates issues with powder hang-up
  • Flat base for a secure, firm rest on the balance pan
  • Supplied nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flat
Black
Square
Antistatic Weighing Dish
44 mm
7 mL
Polystyrene
Non-sterile
44 mm