Learn More
DWK Life Sciences STERIPLAN™ Petri Dish, from Soda-lime Glass
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Brand:  DWK Life Sciences 1184071
17.11 EUR valid until 2024-12-31
Use promo code "21955" to get your promotional price.
 View more versions of this product
Product Code. 11688683
  • 19.80 € / Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow.
  • Free from bubbles and streaks
  • Even distribution of sewing bottom possible
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture
15 mm
Soda Lime Glass
10/Pk.
90 mm