missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Food Scoop
BIOPHARMAGREENER_CHOICE

Can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings.

Brand:  Buerkle™ 5378-5015

 View more versions of this product

Product Code. 17260604

  • 35.95 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Shovel
Not Autoclavable
50 mL
Sterile
Complies with EU food rrequirements and FDA regulations
300 mm
Polystyrene
Long-handled
Single-use
Blue
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ SteriPlast™ Blue Food Scoop > 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.