Learn More
Buerkle™ SteriPlast™ Sampling Spoons
BIOPHARMAGREENER_CHOICE
Sample powders, granules, and pastes. Buerkle™ SteriPlast™ Sampling Spoons are made of polystyrene and come sterile and individually wrapped.
58.40 € - 63.30 €
Specifications
Product Type Spoon
Material Polystyrene
Autoclavable Not Autoclavable
Sterility Sterile
Color White
View More Specs
Products 2
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
15356327
BIOPHARMAGREENER_CHOICE
View Documents
Buerkle™
5378-1011
2.5 mL 127 mm
63.30 €
Pack of 100
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
15351766
BIOPHARMAGREENER_CHOICE
View Documents
Buerkle™
5378-1012
10 mL 170 mm
58.40 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Intended for sampling of powders, granules, and pastes
  • Sterilized by gamma radiation
  • Individually packaged for one-way needs
  • Production, assembling, and packaging according to cleanroom class 7 (10.000)
Specifications
Specifications
Spoon
Not Autoclavable
White
Polystyrene
Sterile
Sampling Powders, Granules, and Pastes