missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SteriSEQ™ Rapid Sterility Testing Kit

The SteriSEQ Rapid Sterility Testing Kit is a TaqMan-based quantitative PCR (qPCR) kit for the detection of bacteria and fungi in complex bioproduction samples.

2373.00 € - 3801.00 €

Specifications

Content And Storage • SteriSEQ™ Assay Mix, 2 x 110 μL; Store at -25°C to -15°C and protect from light
• qPCR Master Mix Plus, 2 x 750 μL; Store at -25°C to -15°C until first thawed, then store at 2°C to 8°C and protect from light
• SteriSEQ™ Discriminatory Positive Control, 2 x 30 μL; Store at -25°C to -15°C
• SteriSEQ™ DNA Dilution Buffer, 1 x 7 mL; Store at -25°C to -15°C until first thawed, then store at room temperature
Detection Method qPCR
Form Liquid
PCR Method qPCR
Label or Dye FAM + VIC, ABY, Alexa Fluor 647
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
18755525
View Documents
Applied Biosystems™
A57186
50 Reactions
2373.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18715415
View Documents
Applied Biosystems™
A57185
100 Reactions
3801.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SteriSEQ Rapid Sterility Testing Kit is a TaqMan-based quantitative PCR (qPCR) kit for the detection of bacteria and fungi in complex bioproduction samples. Designed and tested using criteria for rapid bacteria and fungi detection in biotherapeutic manufacturing cell culture lots, the kit meets or exceeds sensitivity and specificity guidance provided in European Pharmacopoeia (E.P 2.6.27) and U.S. Pharmacopoeia (USP1071). The kit can detect 5–25 genome copies per reaction or the equivalent of 5–25 CFU per reaction.

Features of the SteriSEQ Rapid Sterility Testing Kit include:

  • Fast—sample-to-answer turnaround time of less than a day; delivers results in <5 hours
  • Specific—primers/probes designed specifically for bacteria (16S rRNA) and fungi (18S rRNA)
  • Sensitive—detects bacterial and fungal species at 5–25 genome copies per reaction
  • Efficient—minimizes use of sample material by simultaneously testing for bacteria and fungi, preserving precious cells for final product
  • Accurate—discriminatory positive control helps eliminate false positives and an internal positive control helps ensure PCR reaction consistency in the samples

The SteriSEQ Rapid Sterility Testing Kit uses predefined experiment templates that are included with the following versions of AccuSEQ Real‐Time PCR Software for PCR set up, operation, and post-run analysis:

  • AccuSEQ Real‐Time PCR Software 3.2.1 or later (used with the QuantStudio 5 Real‐Time PCR System)
  • AccuSEQ Real‐Time PCR Software 2.2.1 or later (used with the 7500 Fast Real‐Time PCR Instrument)
Specifications

Specifications

• SteriSEQ™ Assay Mix, 2 x 110 μL; Store at -25°C to -15°C and protect from light
• qPCR Master Mix Plus, 2 x 750 μL; Store at -25°C to -15°C until first thawed, then store at 2°C to 8°C and protect from light
• SteriSEQ™ Discriminatory Positive Control, 2 x 30 μL; Store at -25°C to -15°C
• SteriSEQ™ DNA Dilution Buffer, 1 x 7 mL; Store at -25°C to -15°C until first thawed, then store at room temperature
Liquid
FAM + VIC, ABY, Alexa Fluor 647
Real Time PCR (qPCR)
qPCR
qPCR
SteriSEQ™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.