Learn More
Merck Millipore Steritest™ NEO Device, Green Base for Solvents
Green-base assembly with nylon canister, Durapore™ PVDF membrane, unique drainage design and reinforced base provides increased chemical compatibility for solvents, creams, ointments, and veterinary injectables. Used for sterility testing of products dissolved in solvents in vials or ampoules.
Brand:  Merck Millipore TZHVSL210
Product Code. 15612764
Description
Description
Sterility testing is one of the most crucial and time-consuming steps in pharmaceutical processing. Although it is a critical step, it does not have to be a difficult one. The Steritest™ canister device simplifies every aspect of testing, from handling to traceability to reliability, all within a closed concept system. The ease and convenience enables increases productivity while maintaining the highest levels of quality.
  • Color coded: Green packaging label and device base for easy identification of units designed for testing of products dissolved in solvents
  • Minimize false positives: Closed devices have no open containers or membrane manipulations, which could increase the risk of contamination
  • Reduce false negatives: Eliminates bacteriostatic, fungistatic or bactericidal agents through specific membranes, sealing technology and optimized design
  • Quality check: Rigorous in-process and release quality checks including 100% membrane and canister integrity tests as well as intense physical and microbiological testing
  • Colored Clamps for Accurate Media Filling: Pre-installed clamps and blackline prevent filling errors and improve workflow clarity
  • Volume Graduation: Be precise and improve workflow accuracy through engraved graduation lines at 50, 75 and 100 mL