Learn More
Sampling Systems™ SteriWare™ Cleaning Coupons
BIOPHARMA
SteriWare™ Cleaning Coupons are essential equipment for checking your cleaning processes. Cleaning coupons are available in a range of different materials and two standard sizes.
Brand:  Sampling Systems™ 8352PP-50S
 View more versions of this product
Product Code. 17584646
  • 104.00 € / Pack of 50
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The SteriWare™ Cleaning coupons are the perfect way to test and validate your cleaning regime.
  • The cleanroom production improves the purity of the final product.
  • Each coupon is 100% inspected and then individually bagged within the cleanroom.
  • Sterile coupons are also available from stock.
  • The coupons are available from stock in the popular sizes, materials and finishes.
  • Supplied with Certificate of Conformity for validation purposes.
  • Popular cleaning coupons include PP, nylon, PS, PVC and HDPE.
  • There are a wide range of other coupons available.
  • Made from high quality virgin material.
  • Individually bagged within the class 10,000 cleanroom.
  • Available with batch specific certificate of conformity.
  • Sterile versions are available with an irradiation certificate.
Specifications
Specifications
Polypropylene
Individually Bagged
50 mm
50 mm
Sterile