Learn More
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette is both quick and easy to use and is ideal for taking samples from low viscosity liquids.
Brand: Sampling Systems™ 8051H-1100
This item is not returnable.
View return policy
Description
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Highly visible color coded handle
- Press thumb onto blue end of the tube until lowered to the correct sampling depth
- At the required depth raise thumb until levels equalize
- Press the thumb onto tube to seal in the sample
- No cross contamination
- No cleaning required
Specifications
HDPE | |
225 mL | |
21 mm |
Individually Bagged | |
Non-sterile | |
1100 mm |