Learn More
Sampling Systems™ SteriWare™ Disposable LiquiThief
BIOPHARMA
Sampling Systems™ SteriWare™ Disposable LiquiThief ideal for use with low viscocity liquids.
Brand:  Sampling Systems™ 8050H-1500
Product Code. 15714758
  • 352.00 € / Pack of 20
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • At the required depth pull up the handle
  • The further the handle is pulled, the larger the sample volume
  • Push handle down to expel the sample
  • No cross contamination
  • No cleaning required
Specifications
Specifications
290 mL
Non-sterile, Individually Bagged
20
21 mm
1500 mm