Learn More
Sampling Systems™ SteriWare™ Disposable Powder Funnel
BIOPHARMA
Constructed of chemically resistant, food grade HDPE. Sampling Systems™ SteriWare™ Disposable Powder Funnel is ideal for transferring and sampling powders.
Brand:  Sampling Systems™ 8111H-200
 View more versions of this product
Product Code. 15859471
  • 51.30 € / Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
200 mm
HDPE
Non-sterile
40 mm
10 Pk.