Learn More
Sampling Systems™ SteriWare™ Disposable Slot Sampler
BIOPHARMA
Designed to take a single composite from the top, middle and bottom of your powder bed. Sampling Systems™ SteriWare™ Disposable Slot Samplers are ideal for taking a multilevel, cross sectional sample from powders and granules.
Brand:  Sampling Systems™ 8073J-1000
 View more versions of this product
Product Code. 15809461
  • 305.00 € / Pack of 20
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Push the sampler into the product, ensuring the sampler is in the closed position
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler
  • Deposit the sample into a suitable container
  • No cross contaminatin
  • No cleaning required
Specifications
Specifications
Disposable Slot Sampler
20 per pack
Non-sterile
1000 mm
HDPE
500 mL
40 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number110319 Certificate TypeCertificate of Compliance Date09/10/2023 Catalog Number8073J-1000
Lot Number109519 Certificate TypeCertificate of Compliance Date05/06/2023 Catalog Number8073J-1000
Lot Number103233 Certificate TypeDeclaration of Conformity Date05/12/2022 Catalog Number8073J-1000
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Slot Sampler > Sterility: Non-Sterile