missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable ViscoThief
BIOPHARMA

Sampling Systems™ SteriWare™ Disposable ViscoThief is a single use, disposable sampler that is designed for use with viscous materials such as creams, oils, and honey.

Brand:  Sampling Systems™ 8055P-1000

Product Code. 11919206

  • 190.00 € / Pack of 20
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • At the required depth pull up the handle
  • The further the handle is pulled, the larger the sample volume
  • Push handle down to expel the sample
  • No cross contamination
  • No cleaning required
Specifications

Specifications

Liquid Sampler
Individually bagged
Single-use
21 mm
Polypropylene
190 mL
Non-sterile
1000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado