Learn More
Sampling Systems™ SteriWare™ Disposable Volumetric Scoop
Capable of being used straight from the bag with its individually bagged design. Sampling Systems™ SteriWare™ Disposable Volumetric Scoop allows small volumes to be accurately measured.
63.65 € - 79.10 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Sterility | Price | Quantity & Availability | ||
12734948
|
Sampling Systems™
8640H-01S |
1 mL | 18 mm | 153 mm | Sterile |
78.85 €
Pack of 100 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||
12744948
|
Sampling Systems™
8640H-05S |
5 mL | 29 mm | 165 mm | Sterile |
78.85 €
Pack of 100 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||
12754948
|
Sampling Systems™
8640H-10S |
10 mL | 35 mm | 171 mm | Sterile |
78.85 €
Pack of 100 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||
12764948
|
Sampling Systems™
8640H-20S |
20 mL | 43 mm | 180 mm | Sterile |
79.10 €
Pack of 100 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
||
15819541
|
Sampling Systems™
8640H-01 |
1 mL | 18 mm | 153 mm | Non-sterile |
63.65 €
Pack of 100 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||
15829541
|
Sampling Systems™
8640H-0123S |
1.23 mL | 18 mm | 155 mm | Sterile |
78.85 €
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||
15839541
|
Sampling Systems™
8640H-0246S |
2.46 mL | 23 mm | 160 mm | Sterile |
78.85 €
Pack of 100 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
15849541
|
Sampling Systems™
8640H-05 |
5 mL | 29 mm | 165 mm | Non-sterile |
63.65 €
Pack of 100 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||
15859541
|
Sampling Systems™
8640H-10 |
10 mL | 35 mm | 171 mm | Non-sterile |
63.65 €
Pack of 100 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
15869541
|
Sampling Systems™
8640H-20 |
20 mL | 43 mm | 180 mm | Non-sterile |
63.65 €
Pack of 100 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Available in a variety of sizes
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contaminaton
- No cleaning required
Specifications
Volumetric Scoop | |
HDPE | |
Single-use |
These Volumetric Spoons are not certified measuring devices. User verification required.