Learn More
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief
BIOPHARMA
Ideal for taking representative samples from known locations within the bulk powder. Powder is trapped within the sampler and easily withdrawn and emptied.
Brand:  Sampling Systems™ 8060H-510S
 View more versions of this product
Product Code. 11945636
  • 198.00 € / Pack of 20
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Key Features:
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Available in a variety of lengths and tip sizes
Operation:
  • Insert the sampler into the product ensuring the tip is inside the sampler body.
  • At the required depth pull up the body to expose the tip. The sample will fall in around the tip.
  • Push down body of the sampler to trap the sample.
  • Remove sampler from the product.
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Specifications
Specifications
Powder Thief
HDPE
Single-use
Conforms to FDA CFR 177.1520, EU Regulations 10/2011, EC Regulations 1935/2004
500 mm
HDPE
10 mL
Sterile
Ideal for taking representative samples from known locations within the bulk powder