Learn More
Sampling Systems™ SteriWare™ Mono Slot Sampler
BIOPHARMA
Designed to take a 50mL sample from the sampling slot positioned near the tip of the sampler. Sampling Systems™ SteriWare™ Disposable Mono Slot Sampler is ideal for taking a single sample from free flowing powders and granules.
Brand:  Sampling Systems™ 8069H-500
 View more versions of this product
Product Code. 15879451
  • 178.00 € / Pack of 20
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Push the sampler into the product, ensuring the sampler is in the closed position
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler
  • Deposit the sample into a suitable container
  • No cross contaminatin
  • No cleaning required
Specifications
Specifications
Slot Sampler
20 per pack
Non-sterile
620 mm
HDPE
50 mL
26 mm