Learn More
Sampling Systems™ SteriWare™ Powder Thief
BIOPHARMA
Learn how to prevent cross contamination when sampling when you use a SteriWare™ PowderThief sampler. With a SteriWare™ PowderThief sampler, you can take samples from most free-flowing powders and granules.
Brand:  Sampling Systems™ 8060H-1520S
 View more versions of this product
Product Code. 17554596
  • 360.00 € / Pack of 20
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The sterile, disposable PowderThief is ideal for taking representative samples from known locations within the bulk powder.
  • These samplers are available in two convenient lengths and four sample volumes.
  • Disposable Powder Thief samplers are available sterile or non-sterile.
  • Assembled and packed in a cleanroom.
  • Individually bagged.
  • Designed for single use.
  • Made from FDA and EU conforming HDPE.
  • BSE/TSE Free.
  • Full Batch Traceability.
  • Available as gamma irradiated sterile (SAL 10-6).
Specifications
Specifications
HDPE
Single-use
Individually Bagged
20 mL
Sterile
1500 mm