Learn More
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™
BIOPHARMA
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Brand:  Sampling Systems™ 8045R-250S
 View more versions of this product
Product Code. 17498873
  • 209.00 € / Pack of 85
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
Disposable Scoop
Individually Bagged
Single-use
Red
Polystyrene
250 mL
Sterile