Learn More
Sampling Systems™ SteriWare™ Sterile HDPE Disposable Powder Thief
BIOPHARMA
Sampling Systems™ SteriWare™ Sterile HDPE Disposable Powder Thief is ideal for taking representative samples from known locations within the bulk powder.
156.00 € - 259.00 €
Specifications
Material HDPE
Sterility Sterile
Diameter (Metric) 21 mm
View More Specs
Products 7
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
11965437
BIOPHARMA
View Documents
Sampling Systems™
8060H-501S
1 mL 500 mm
156.00 €
Pack of 20
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
11925457
BIOPHARMA
View Documents
Sampling Systems™
8060H-1001S
1 mL 1000 mm
209.00 €
Pack of 20
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
11925636
BIOPHARMA
View Documents
Sampling Systems™
8060H-1005S
5 mL 1000 mm
209.00 €
Pack of 20
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
11965636
BIOPHARMA
View Documents
Sampling Systems™
8060H-1010S
10 mL 1000 mm
209.00 €
Pack of 20
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
11995437
BIOPHARMA
View Documents
Sampling Systems™
8060H-520S
20 mL 500 mm
156.00 €
Pack of 20
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
11905636
BIOPHARMA
View Documents
Sampling Systems™
8060H-505S
5 mL 500 mm
156.00 €
Pack of 20
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
11945457
BIOPHARMA
View Documents
Sampling Systems™
8060H-1020S
20 mL 1000 mm
259.00 €
Pack of 20
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Description
Description
  • Assembled and packed in a cleanroom
  • Individually Bagged
  • Made from FDA or EU conforming HDPE
  • Available as gamma irradiated sterile (SAL 10-6)
  • BSE/TSE Free
  • Full Batch Traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • No cleaning validation (this is very expensive to set up and run)
  • Manufactured in a class 100,000 cleanroom environment (check individual sampler descriptions)
  • Peace of mind
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Financial
  • Save time and money (it will cost more to clean a reusable sampler than to buy a disposable sampler)
Specifications
Specifications
HDPE
Sterile
21 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Sterile HDPE Disposable Powder Thief