missing translation for 'onlineSavingsMsg'
Learn More

Stigmasterol, 95%, Thermo Scientific Chemicals

Product Code. 10274750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10274750

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-48-7
Molecular Formula C29H48O
Molecular Weight (g/mol) 412.70
MDL Number MFCD00003630
InChI Key HCXVJBMSMIARIN-PHZDYDNGSA-N
Synonym stigmasterol, stigmasterin, beta-stigmasterol, stigmasta-5,22-dien-3beta-ol, 24s-5,22-stigmastadien-3beta-ol, unii-99wuk5d0y8, stigmasta-5,22e-dien-3beta-ol, stigmasta-5,22-dien-3-beta-ol, 3beta,22e-stigmasta-5,22-dien-3-ol, stigmasta-5,22-dien-3-ol, 3b,22e
PubChem CID 5280794
ChEBI CHEBI:28824
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Melting Point 161.0°C to 170.0°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 8944
Specific Rotation Condition - 50.00 (20.00°C c=2,CHCl3)
Specific Rotation − 50.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 412.7
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Stigmasterol, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.