Learn More
Cole-Parmer™ Stir Bars
Stir Bars suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
Brand:  Cole-Parmer™ 36630-99
 View more versions of this product
Product Code. 12799038
  • 77.85 € / Pack of 10
Estimated Shipment: 09-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Specifications
Specifications
0.18 in.
Integrity 10, RS-200, and RS-250 Reaction Stations
4.5 mm
Cylinder
For Stem Integrity
0.5 in.
12 mm
Magnetic Stir Bar