Learn More
Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal are ideal for use with applications under vacuum or under slight overpressure.
Brand:  Lenz Laborglasinstrumente™ 06306206
 View more versions of this product
Product Code. 11913931
  • 443.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With extended guidance for reduced vibrations and a longer lifetime
  • Made of borosilicate glass
  • PTFE(sealings)
Specifications
Specifications
16 mm
Borosilicate Glass
Joint (NS) 45/46
PTFE