Learn More
Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal are ideal for use with applications under vacuum or under slight overpressure.
Brand: Lenz Laborglasinstrumente™ 06306204
This item is not returnable.
View return policy
Description
- With extended guidance for reduced vibrations and a longer lifetime
- Made of borosilicate glass
- PTFE(sealings)
Specifications
16 mm | |
Borosilicate Glass | |
Joint (NS) 29/32 | |
PTFE |