Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
Brand:  SP Bel-Art™ F37110-1608
 View more versions of this product
Product Code. 16448485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • With pivot ring for better mixing conditions
  • Alnico magnet encapsulated with PTFE™ brand PTFE coating
  • FDA Grade, USP Class VI
  • Good chemical compatibility
  • Autoclavable
Specifications
Specifications
8 mm
16 mm
10/Pk.
Octagon