Learn More
Novus Biologicals™ Stomach Tissue Slides (Adenocarcinoma)- Paraffin
Brand:  Novus Biologicals™ NBP2-30309
Product Code. 18125912
  • 207.00 € / Pack of 5
Explore more special offers
Description
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
Human
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Adenocarcinoma
Stomach Tissue Slides (Adenocarcinoma)