Learn More
Thermo Fisher Scientific™ FermionX Stomacher™ 400 Bag Opener
The Stomacher™ 400 bag opener from FermionX holds open bags for easy addition of samples without risking instrument contact on the bag.
Brand:  Thermo Fisher Scientific™ BA6095
Product Code. 12764876
  • 54.75 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Stomacher™ 400 bag opener from FermionX holds open bags for easy addition of samples without risking instrument contact on the bag.
  • Compatible with Stomacher™ 80 and Stomacher™ 400 bag sizes
  • Stable on benchtop or scales
  • Easily cleaned with most lab disinfectant
  • Capacity: One place
  • Dimensions (L x W x H): 15.5 x 11.5 x 27 cm
Specifications
Specifications
Bag Opener
Plastic polymer
Stomacher™ 80 and Stomacher™ 400 bag
1