Learn More
Buerkle™ Stop-It™ Hose Clamp with Easy-Click Closure
Regulate the flow of liquids and gasses with these hose clips. Buerkle™ Easy-Click™ Closure features a knurled screw for stepless, delicate flow regulation or complete stop of the flow. The closure compresses the hose over a wide area against the steel hook to prevent damage to the hose.
Brand:  Buerkle™ 8619-0106
Product Code. 10526092
  • 5.50 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Easy-Click closure - stop-it can be opened and closed in any position - without tiresome screwing on and off. High quality material - stop-it made from the heat resistant and chemical resistant plastic PVDF. The clamp is galvanised steel.
Specifications
Specifications
Easy Click Closure
10 mm Flexible Hose
50/Scale
10 mm
160°C
PVDF and Steel (Zinc-coated)
It Can be Used Wherever The Flow in Hoses with Liquids or Gases Needs to be Regulated or Stopped
10 mm
Green