missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper
Provide airtight covers
Brand: DWK Life Sciences 224100-180
324.32 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
2 cm | |
Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials | |
Autoclavable | |
13 mm | |
Plug Stopper |
20 mm | |
45 Durometer | |
Red | |
Rubber |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction