Learn More
BRAND™ Storage Box For Cryogenic Tubes
Holds 81 tubes
Brand:  BRAND™ 114864
 View more versions of this product
Product Code. 15501281
  • 146.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polycarbonate (PC)
  • For cryogenic tubes
  • Operating range (in the gaseous phase of liquid nitrogen) -196 °C to +121 °C
  • Autoclavable at 121 °C (2 bar)
  • DIN EN 285
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Polycarbonate
    Cryo Storage Box
    81 (3, 4 and 5 mL Cryogenic Tubes)
    5