Learn More
Fisherbrand™ Tubby™ Storage Containers
GREENER_CHOICE
Store, organize and transport small items to maintain a tidy lab. Fisherbrand™ Tubby™ Storage Containers feature removable dividers for customized bin compartments. Features tight-fitting lids and heavy-impact, clear polypropylene.
Brand:  Fisherbrand™ HS23453
Product Code. 11938014
  • 45.28 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Stackable and proportionately sized to utilize space efficiently
  • Removable dividers create as big or as small a compartment as needed
  • Lid snaps in place to secure the contents for safe transport
  • Clear sides for at-a-glance access and inventory monitoring
  • Ideal for tubes, tips, pipettes, gloves and other small items
  • 5-pack includes 5 tubs, 5 lids and 15 dividers
Specifications
Specifications
Storage Bin
Clear
5/Pk.
200 mm
Polypropylene
Divider, Lid
330 mm
115 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Tubby™ Storage Containers > Dimensions: 200L x 330W x 115mmH